Fumarsäuremonoethylester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(E)-4-Ethoxy-4-oxobut-2-enoic acid, Monoethyl fumarate, Ethyl hydrogen fumarate
M-Clarity™ Qualitätsstufe = MQ100
128422-25G
SIAL128422-25GEA
66.9
EUR
SIAL128422-25G
Fumarsäuremonoethylester, Sigma-Aldrich®
Fumarsäuremonoethylester
Formel:
C₆H₈O₄ Molecular Weight: 144,13 g/mol |
MDL:
MFCD00002699 CAS-Nummer: 2459-05-4 EINECS: 219-544-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...