trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

128708-100G 128708-25G 128708-5G
SIAL128708-100GEA 147 EUR
SIAL128708-100G SIAL128708-25G SIAL128708-5G
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, Sigma-Aldrich®
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid
Formel: C₁₀H₁₀O₄
MDL: MFCD00004400
CAS-Nummer: 537-98-4
EINECS: 208-679-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR