1-Chlor-3,5-dibrombenzol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 3,5-Dibromochlorobenzene, 1,3-Dibromo-5-chlorobenzene

M-Clarity™ Qualitätsstufe = MQ100

129127-5G
SIAL129127-5GEA 28.5 EUR
SIAL129127-5G
1-Chlor-3,5-dibrombenzol, Sigma-Aldrich®
1-Chloro-3,5-dibromobenzene
Formel: C₆H₃Br₂Cl
Molecular Weight: 270,35 g/mol
MDL: MFCD00070765
CAS-Nummer: 14862-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR