Magnesiumacetylacetonat Dihydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(2,4-pentandionato-O,O')magnesium Dihydrat, Magnesium-2,4-pentandionat Dihydrat, Magnesiumbis(2,4-pentandionato-O,O') Dihydrat

M-Clarity™ Qualitätsstufe = MQ100

129577-25G
SIAL129577-25GEA 113 EUR
SIAL129577-25G
Magnesiumacetylacetonat Dihydrat, Sigma-Aldrich®
Magnesiumacetylacetonat Dihydrat
Formel: C₁₀H₁₄MgO₄·2H₂O
Molecular Weight: 258,55 g/mol
MDL: MFCD00150168
CAS-Nummer: 68488-07-3
EINECS: 237-857-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR