Methylcyclopentadien dimer, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

129828-500ML
SIAL129828-500MLEA 185 EUR
SIAL129828-500ML
Methylcyclopentadien dimer, Sigma-Aldrich®
Methylcyclopentadien dimer
Formel: C₁₂H₁₆
MDL: MFCD00082297
CAS-Nummer: 26472-00-4
EINECS: 247-724-5
UN: 3295
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR