Methyl-2-furoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2-Furancarboxylate, Methyl furan-2-carboxylate, Methyl 2-furoate

M-Clarity™ Qualitätsstufe = MQ100

129852-25G
SIAL129852-25GEA 41.2 EUR
SIAL129852-25G
Methyl-2-furoat, Sigma-Aldrich®
Methyl-2-furoat
Formel: C₆H₆O₃
Molecular Weight: 126,11 g/mol
MDL: MFCD00003236
CAS-Nummer: 611-13-2
EINECS: 210-254-6
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR