(±)-2-Methylglutarsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methylpentanedioic acid

M-Clarity™ Qualitätsstufe = MQ100

129860-5G
SIAL129860-5GEA 67 EUR
SIAL129860-5G
(±)-2-Methylglutarsäure, Sigma-Aldrich®
(±)-2-Methylglutarsäure
Formel: C₆H₁₀O₄
Molecular Weight: 146,14 g/mol
MDL: MFCD00002661
CAS-Nummer: 617-62-9
EINECS: 210-521-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR