(2-Nitrophenyl)essigsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
130249-25G
SIAL130249-25GEA
84.9
EUR
SIAL130249-25G
(2-Nitrophenyl)essigsäure, Sigma-Aldrich®
(2-Nitrophenyl)essigsäure
Formel:
(C₆H₄NO₂)CH₂COOH Schmelzpunkt: 136…139 °C |
MDL:
MFCD00007190 CAS-Nummer: 3740-52-1 EINECS: 223-128-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...