5,7-Dichlor-2-methyl-8-chinolinol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5,7-Dichloro-8-hydroxy-2-methylquinoline, 5,7-Dichloro-2-methylquinolin-8-ol
M-Clarity™ Qualitätsstufe = MQ200
130532-10G
SIAL130532-10GEA
54.5
EUR
SIAL130532-10G
5,7-Dichlor-2-methyl-8-chinolinol, Sigma-Aldrich®
5,7-Dichlor-2-methyl-8-chinolinol
Formel:
C₁₀H₇Cl₂NO Molecular Weight: 228,0747 g/mol |
MDL:
MFCD00023984 CAS-Nummer: 72-80-0 EINECS: 200-789-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...