3'-Nitropropiophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: m-Nitropropiophenon

M-Clarity™ Qualitätsstufe = MQ100

130761-5G
SIAL130761-5GEA 66.1 EUR
SIAL130761-5G
3'-Nitropropiophenon, Sigma-Aldrich®
3'-Nitropropiophenon
Formel: C₉H₉NO₃
Molecular Weight: 179,18 g/mol
MDL: MFCD00051516
CAS-Nummer: 17408-16-1
EINECS: 241-435-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR