(±)-α-Chlorphenylacetylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

130966-10G
SIAL130966-10GEA 275 EUR
SIAL130966-10G
(±)-α-Chlorphenylacetylchlorid, Sigma-Aldrich®
(±)-α-Chlorphenylacetylchlorid
Formel: C₈H₆Cl₂O
Molecular Weight: 189,04 g/mol
CAS-Nummer: 2912-62-1
EINECS: 220-826-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR