1,2-Phenylendiessigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: o-Phenylenediacetic acid, 2,2'-(1,2-Phenylene)diacetic acid

M-Clarity™ Qualitätsstufe = MQ100

131407-5G
SIAL131407-5GEA 53.9 EUR
SIAL131407-5G
1,2-Phenylendiessigsäure, Sigma-Aldrich®
1,2-Phenylendiessigsäure
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
MDL: MFCD00004329
CAS-Nummer: 7500-53-0
EINECS: 231-355-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR