(1-Propyl)triphenylphosphoniumbromid, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Triphenyl(propyl)phosphonium bromide, Triphenylpropylphosphonium bromide

131563-25G
SIAL131563-25GEA 110 EUR
SIAL131563-25G
(1-Propyl)triphenylphosphoniumbromid, Sigma-Aldrich®
(1-Propyl)triphenylphosphoniumbromid
Formel: C₂₁H₂₂BrP
Molecular Weight: 385,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011843
CAS-Nummer: 6228-47-3
EINECS: 228-330-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR