Mangan(II)chlorid Monohydrat, Sigma-Aldrich®

Lieferant: Merck
Achtung


MQ200

13220-2.5KG-R 13220.9030
SIAL13220-2.5KG-REA 180 EUR
SIAL13220-2.5KG-R HONE13220.9030
Mangan(II)chlorid Monohydrat, Sigma-Aldrich®
Mangan(II)chlorid Monohydrat
Formel: Cl₂Mn·1H₂O
Molecular Weight: 143.86 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 64333-01-3
EINECS: 231-869-6
UN: 3077
ADR: 9 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR