Diethyl(3-chlorpropyl)malonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diethyl 2-(3-chloropropyl)malonate

M-Clarity™ Qualitätsstufe = MQ100

132446-5G
SIAL132446-5GEA 60.4 EUR
SIAL132446-5G
Diethyl(3-chlorpropyl)malonat, Sigma-Aldrich®
Diethyl(3-chlorpropyl)malonat
Formel: C₁₀H₁₇ClO₄
Molecular Weight: 236,70 g/mol
MDL: MFCD00009169
CAS-Nummer: 18719-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR