Pyridiniumtribromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pyridinium bromide perbromide, Monopyridin-1-ium tribromide, Pyridine hydrobromide perbromide

M-Clarity™ Qualitätsstufe = MQ200

133248-100G 133248-25G 133248-500G
SIAL133248-100GEA 80.9 EUR
SIAL133248-100G SIAL133248-25G SIAL133248-500G
Pyridiniumtribromid, Sigma-Aldrich®
Pyridinium tribromide
Formel: C₅H₆Br₃N
Molecular Weight: 319.82 g/mol
MDL: MFCD00013223
CAS-Nummer: 39416-48-3
EINECS: 254-446-8
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR