1-Amino-2-naphthol Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

133477-5G
SIAL133477-5GEA 219 EUR
SIAL133477-5G
1-Amino-2-naphthol Hydrochlorid, Sigma-Aldrich®
1-Amino-2-naphthol Hydrochlorid
Formel: C₁₀H₁₀ClNO
Molecular Weight: 195,65 g/mol
MDL: MFCD00012561
CAS-Nummer: 1198-27-2
EINECS: 214-830-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR