Benzyldimethyldodecylammoniumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzyldodecyldimethylammonium bromide, Benzalkonium bromide, N-Benzyl-N,N-dimethyldodecan-1-aminium bromide

13373-10G-F 13373-50G-F
SIAL13373-10G-FEA 254 EUR
SIAL13373-10G-F SIAL13373-50G-F
Benzyldimethyldodecylammoniumbromid, Sigma-Aldrich®
Benzyldimethyldodecylammoniumbromid
Formel: C₂₁H₃₈BrN
Molecular Weight: 384.44 g/mol
MDL: MFCD00011768
CAS-Nummer: 7281-04-1
EINECS: 230-698-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR