trans-2-Phenylcyclopropancarbonylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

134309-5G
SIAL134309-5GEA 226 EUR
SIAL134309-5G
trans-2-Phenylcyclopropancarbonylchlorid, Sigma-Aldrich®
trans-2-Phenylcyclopropancarbonylchlorid
Formel: C₁₀H₉ClO
CAS-Nummer: 939-87-7
EINECS: 213-365-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR