5-Chlormethyl-2-oxazolidinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-(Chloromethyl)oxazolidin-2-one

M-Clarity™ Qualitätsstufe = MQ100

135658-5G
SIAL135658-5GEA 73.2 EUR
SIAL135658-5G
5-Chlormethyl-2-oxazolidinon, Sigma-Aldrich®
5-Chlormethyl-2-oxazolidinon
Formel: C₄H₆ClNO₂
Molecular Weight: 135,55 g/mol
MDL: MFCD00005269
CAS-Nummer: 22625-57-6
EINECS: 245-137-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR