Chinuclidin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Azabicyclo[2.2.2]octane hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

135917-1G
SIAL135917-1GEA 114 EUR
SIAL135917-1G
Chinuclidin Hydrochlorid, Sigma-Aldrich®
Chinuclidin Hydrochlorid
Formel: C₇H₁₄ClN
Molecular Weight: 147.650 g/mol
MDL: MFCD00012727
CAS-Nummer: 39896-06-5
EINECS: 254-682-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR