3,3-Diphenylpropylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,3-Diphenylpropan-1-amine

M-Clarity™ Qualitätsstufe = MQ100

136298-25G
SIAL136298-25GEA 71.9 EUR
SIAL136298-25G
3,3-Diphenylpropylamin, Sigma-Aldrich®
3,3-Diphenylpropylamin
Formel: C₁₅H₁₇N
Molecular Weight: 211,31 g/mol
MDL: MFCD00008202
CAS-Nummer: 5586-73-2
EINECS: 226-984-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR