2-Methyl-2-oxazolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methyl-4,5-dihydrooxazole

M-Clarity™ Qualitätsstufe = MQ100

137448-100G
SIAL137448-100GEA 78.9 EUR
SIAL137448-100G
2-Methyl-2-oxazolin, Sigma-Aldrich®
2-Methyl-2-oxazolin
Formel: C₄H₇NO
Molecular Weight: 85,11 g/mol
MDL: MFCD00005298
CAS-Nummer: 1120-64-5
EINECS: 214-316-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR