2-Amino-4-methoxybenzothiazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Methoxybenzothiazol-2-ylamine
138215-5G
SIAL138215-5GEA
79.1
EUR
SIAL138215-5G
2-Amino-4-methoxybenzothiazol, Sigma-Aldrich®
2-Amino-4-methoxybenzothiazole
Formel:
C₈H₈N₂OS Molecular Weight: 180,23 g/mol Siedepunkt: 335 °C (1013 hPa) Schmelzpunkt: 154 °C |
MDL:
MFCD00005792 CAS-Nummer: 5464-79-9 EINECS: 226-763-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...