N-(2-Hydroxyethyl)phthalimid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(2-Hydroxyethyl)phthalsäureimid, N-(2-Hydroxyethyl)-1,2-benzoldicarbonsäureimid

138339-100G
SIAL138339-100GEA 62.8 EUR
SIAL138339-100G
N-(2-Hydroxyethyl)phthalimid, Sigma-Aldrich®
N-(2-Hydroxyethyl)phthalimid
Formel: C₁₀H₉NO₃
MDL: MFCD00005903
CAS-Nummer: 3891-07-4
EINECS: 223-434-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR