Cyclohexylphenylketon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

139211-25G
SIAL139211-25GEA 104 EUR
SIAL139211-25G
Cyclohexylphenylketon, Sigma-Aldrich®
Cyclohexylphenylketon
Formel: C₁₃H₁₆O
Molecular Weight: 188,27 g/mol
MDL: MFCD00001467
CAS-Nummer: 712-50-5
EINECS: 211-923-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR