Hexafluoraceton Trihydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
139238-25G
139238-5G
SIAL139238-25GEA
73.1
EUR
SIAL139238-25G
SIAL139238-5G
Hexafluoraceton Trihydrat, Sigma-Aldrich®
Hexafluoraceton Trihydrat
Formel:
C₃F₆O·3H₂O |
MDL:
MFCD00149061 CAS-Nummer: 34202-69-2 EINECS: 211-676-3 UN: 2552 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...