2,5-Dimercapto-1,3,4-thiadiazol Dikaliumsalz, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

139432-25G
SIAL139432-25GEA 120 EUR
SIAL139432-25G
2,5-Dimercapto-1,3,4-thiadiazol Dikaliumsalz, Sigma-Aldrich®
2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt
Formel: C₂K₂N₂S₃
Molecular Weight: 226,43 g/mol
MDL: MFCD00003104
CAS-Nummer: 4628-94-8
EINECS: 225-043-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR