1,1'-[Oxybis(ethylenoxy)]diethylen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-(2-Ethenyloxyethoxy)ethoxy)ethen, 1,1'-[Oxybis(ethylenoxy)]diethen, Diethylenglycoldivinylether, 1,1'-(Oxybis(ethylenoxy))diethen, 1,1'-(Oxybis(ethylenoxy))diethylen

139548-50ML
SIAL139548-50MLEA 250 EUR
SIAL139548-50ML
1,1'-[Oxybis(ethylenoxy)]diethylen, Sigma-Aldrich®
1,1'-[Oxybis(ethylenoxy)]diethylen
Formel: H₂CCHO(CH₂)₂O(CH₂)₂OCHCH₂
Molecular Weight: 158,2 g/mol
Dichte: 0,97 g/cm³ (20 °C)
MDL: MFCD00054610
CAS-Nummer: 764-99-8
EINECS: 212-133-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR