4-Hydroxy-3,5-dimethylbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 3,5-Dimethyl-4-hydroxybenzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

140392-5G
SIAL140392-5GEA 50.8 EUR
SIAL140392-5G
4-Hydroxy-3,5-dimethylbenzaldehyd, Sigma-Aldrich®
4-Hydroxy-3,5-dimethylbenzaldehyd
Formel: C₉H₁₀O₂
Molecular Weight: 150,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006946
CAS-Nummer: 2233-18-3
EINECS: 218-774-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR