5-Iodisatin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

140589-25G 140589-5G
SIAL140589-25GEA 86.7 EUR
SIAL140589-25G SIAL140589-5G
5-Iodisatin, Sigma-Aldrich®
5-Iodisatin
Formel: C₈H₄INO₂
MDL: MFCD00016899
CAS-Nummer: 20780-76-1
EINECS: 244-035-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR