Thiophen-2-acetonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Thiophene-2-acetonitrile, 2-Thiopheneacetonitrile, 2-(Thiophen-2-yl)acetonitrile

M-Clarity™ Qualitätsstufe = MQ200

141682-10G
SIAL141682-10GEA 50.6 EUR
SIAL141682-10G
Thiophen-2-acetonitril, Sigma-Aldrich®
Thiophen-2-acetonitril
Formel: C₆H₅NS
Molecular Weight: 123,18 g/mol
MDL: MFCD00005453
CAS-Nummer: 20893-30-5
EINECS: 244-104-6
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR