6-Ethoxy-2-mercaptobenzothiazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Ethoxybenzo[d]thiazole-2(3H)-thione

M-Clarity™ Qualitätsstufe = MQ100

142174-5G
SIAL142174-5GEA 92.3 EUR
SIAL142174-5G
6-Ethoxy-2-mercaptobenzothiazol, Sigma-Aldrich®
6-Ethoxy-2-mercaptobenzothiazole
Formel: C₉H₉NOS₂
Molecular Weight: 211,31 g/mol
MDL: MFCD00467715
CAS-Nummer: 120-53-6
EINECS: 204-405-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR