2-Chlor-6-nitrobenzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Chloro-2-nitrobenzonitrile

142255-5G
SIAL142255-5GEA 72.8 EUR
SIAL142255-5G
2-Chlor-6-nitrobenzonitril, Sigma-Aldrich®
2-Chlor-6-nitrobenzonitril
Formel: C₇H₃ClN₂O₂
Molecular Weight: 182,57 g/mol
MDL: MFCD00007203
CAS-Nummer: 6575-07-1
EINECS: 229-496-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR