3-Ethylpyridin, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
142395-5G
SIAL142395-5GEA
78.6
EUR
SIAL142395-5G
3-Ethylpyridin, Sigma-Aldrich®
3-Ethylpyridine
Formel:
C₇H₉N Molecular Weight: 107.16 g/mol |
MDL:
MFCD00006413 CAS-Nummer: 536-78-7 EINECS: 208-647-2 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...