Betain Monohydrat, Sigma-Aldrich®
Lieferant: Merck
14300-100G
14300-500G
SIAL14300-100GEA
54.4
EUR
SIAL14300-100G
SIAL14300-500G
Betain Monohydrat, Sigma-Aldrich®
Betain Monohydrat
Formel:
C₅H₁₁NO₂·1H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150010 CAS-Nummer: 590-47-6 EINECS: 203-490-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...