trans-2,3-Dibrom-2-buten-1,4-diol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

143707-100G
SIAL143707-100GEA 75.7 EUR
SIAL143707-100G
trans-2,3-Dibrom-2-buten-1,4-diol, Sigma-Aldrich®
trans-2,3-Dibrom-2-buten-1,4-diol
Formel: C₄H₆Br₂O₂
MDL: MFCD00004698
CAS-Nummer: 21285-46-1
EINECS: 244-313-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR