2,4-Dichlorpyrimidin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
MQ200
143847-10G
143847-50G
SIAL143847-10GEA
79.5
EUR
SIAL143847-10G
SIAL143847-50G
2,4-Dichlorpyrimidin, Sigma-Aldrich®
2,4-Dichloropyrimidine
Formel:
C₄H₂Cl₂N₂ Molecular Weight: 148.98 g/mol Siedepunkt: 209 Schmelzpunkt: 61 |
MDL:
MFCD00006061 CAS-Nummer: 3934-20-1 EINECS: 223-508-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...