Diethyldimethylmalonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diethyl 2,2-dimethylmalonate

M-Clarity™ Qualitätsstufe = MQ100

143901-10G
SIAL143901-10GEA 129 EUR
SIAL143901-10G
Diethyldimethylmalonat, Sigma-Aldrich®
Diethyldimethylmalonat
Formel: C₉H₁₆O₄
Molecular Weight: 188,2200 g/mol
MDL: MFCD00009128
CAS-Nummer: 1619-62-1
EINECS: 216-582-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR