2,1-Benzisoxazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Anthranil, Benzo[c]isoxazole, 2,1-Benzisoxazole

M-Clarity™ Qualitätsstufe = MQ100

144517-1G
SIAL144517-1GEA 41.2 EUR
SIAL144517-1G
2,1-Benzisoxazol, Sigma-Aldrich®
2,1-Benzisoxazole
Formel: C₇H₅NO
Molecular Weight: 119,1207 g/mol
MDL: MFCD00005806
CAS-Nummer: 271-58-9
EINECS: 205-980-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR