3,4-Dichlorzimtsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(3,4-Dichlorphenyl)propensäure, 3,4-Dichlorcinnamsäure

M-Clarity™ Qualitätsstufe = MQ100

144703-25G
SIAL144703-25GEA 116 EUR
SIAL144703-25G
3,4-Dichlorzimtsäure, Sigma-Aldrich®
3,4-Dichlorzimtsäure
Formel: C₉H₆Cl₂O₂
Molecular Weight: 217,05 g/mol
MDL: MFCD00004385
CAS-Nummer: 1202-39-7
EINECS: 214-866-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR