Chinoxalin-2,3-dion, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,3-Dihydroxyquinoxaline, Quinoxaline-2,3(1H,4H)-dione
M-Clarity™ Qualitätsstufe = MQ100
144789-25G
SIAL144789-25GEA
35.7
EUR
SIAL144789-25G
Chinoxalin-2,3-dion, Sigma-Aldrich®
Chinoxalin-2,3-dion
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,15 g/mol |
MDL:
MFCD00006723 CAS-Nummer: 15804-19-0 EINECS: 239-901-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...