1-(Trimethylsiloxy)cyclohexen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (Cyclohex-1-en-1-yloxy)trimethylsilane

144819-5G
SIAL144819-5GEA 118 EUR
SIAL144819-5G
1-(Trimethylsiloxy)cyclohexen, Sigma-Aldrich®
1-(Trimethylsiloxy)cyclohexen
Formel: C₉H₁₈OSi
Molecular Weight: 170.33 g/mol
MDL: MFCD00001541
CAS-Nummer: 6651-36-1
EINECS: 229-675-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR