4-Ethoxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

144959-25G
SIAL144959-25GEA 32.5 EUR
SIAL144959-25G
4-Ethoxybenzoesäure, Sigma-Aldrich®
4-Ethoxybenzoic acid
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Schmelzpunkt: 199,6 °C
MDL: MFCD00002545
CAS-Nummer: 619-86-3
EINECS: 210-616-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR