3,4-(Methylendioxy)phenylacetonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

145300-10G
SIAL145300-10GEA 84.7 EUR
SIAL145300-10G
3,4-(Methylendioxy)phenylacetonitril, Sigma-Aldrich®
3,4-(Methylendioxy)phenylacetonitril
Formel: C₉H₇NO₂
Molecular Weight: 161.16 g/mol
MDL: MFCD00005835
CAS-Nummer: 4439-02-5
EINECS: 224-655-9
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR