4-Brom-α-methylbenzylalkohol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Bromophenyl methyl carbinol, 1-(4-Bromophenyl)ethanol, p-Bromo-α-methylbenzyl alcohol
M-Clarity™ Qualitätsstufe = MQ100
145769-5G
SIAL145769-5GEA
31.9
EUR
SIAL145769-5G
4-Brom-α-methylbenzylalkohol, Sigma-Aldrich®
4-Brom-α-methylbenzylalkohol
Formel:
C₈H₉BrO Molecular Weight: 201,06 g/mol Siedepunkt: 253 °C (1013 hPa) Dichte: 1,469 g/cm³ (15.2 °C) |
MDL:
MFCD00004514 CAS-Nummer: 5391-88-8 EINECS: 226-389-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...