Bis(1,5-cyclooctadien)rhodium(I)tetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(cycloocta-1,5-dien)rhodium(I)-tetrafluoroborat

14694-100MG
SIAL14694-100MGEA 144 EUR
SIAL14694-100MG
Bis(1,5-cyclooctadien)rhodium(I)tetrafluoroborat, Sigma-Aldrich®
Bis(1,5-cyclooctadien)rhodium(I)tetrafluoroborat
Formel: C₁₆H₂₄BF₄Rh
Molecular Weight: 406,072 g/mol
MDL: MFCD00075045
CAS-Nummer: 35138-22-8
EINECS: 460-220-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR