N-Ethylacetamid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

147400-25G
SIAL147400-25GEA 86.8 EUR
SIAL147400-25G
N-Ethylacetamid, Sigma-Aldrich®
N-Ethylacetamid
Formel: C₄H₉NO
Molecular Weight: 87,12 g/mol
MDL: MFCD00009029
CAS-Nummer: 625-50-3
EINECS: 210-896-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR