4-Chlor-α-methylbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(4-Chlorophenyl)ethanol, 4-Chlorophenyl methyl carbinol, p-Chloro-α-methylbenzyl alcohol

M-Clarity™ Qualitätsstufe = MQ100

147559-25G
SIAL147559-25GEA 78 EUR
SIAL147559-25G
4-Chlor-α-methylbenzylalkohol, Sigma-Aldrich®
4-Chlor-α-methylbenzylalkohol
Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
Dichte: 1,17 g/cm³ (20 °C)
MDL: MFCD00041038
CAS-Nummer: 3391-10-4
EINECS: 222-223-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR