Diethylcyclopropan-1,1-dicarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

147710-25G
SIAL147710-25GEA 317 EUR
SIAL147710-25G
Diethylcyclopropan-1,1-dicarboxylat, Sigma-Aldrich®
Diethylcyclopropan-1,1-dicarboxylat
Formel: C₉H₁₄O₄
MDL: MFCD00001283
CAS-Nummer: 1559-02-0
EINECS: 216-321-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR